missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£161.00 - £367.00
Specifications
| Antigen | BP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18040284
|
Novus Biologicals
NB100-481SS |
0.025 mL |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087563
|
Novus Biologicals
NB100-481 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BP1 Polyclonal specifically detects BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| BP1 | |
| Western Blot 1:1000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8 | |
| DLX4 | |
| IgG | |
| Affinity Purified | |
| This is specific for human BP1 (DLX4 variant 1). Berg PE, Kirolikar S. DLX4 (distal-less homeobox 4). Atlas Genet Cytogenet Oncol Haematol. January 2011. URL : http://AtlasGeneticsOncology.org/Genes/DLX4ID49827ch17q21.html |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP Assay | |
| Unconjugated | |
| RUO | |
| Human | |
| Q92988 | |
| 1748 | |
| A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt# Q92988]. | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 36 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title