missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BP1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-481V
This item is not returnable.
View return policy
Description
BP1 Polyclonal specifically detects BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| BP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Q92988 | |
| DLX4 | |
| A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt# Q92988]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for human BP1 (DLX4 variant 1). Berg PE, Kirolikar S. DLX4 (distal-less homeobox 4). Atlas Genet Cytogenet Oncol Haematol. January 2011. URL : http://AtlasGeneticsOncology.org/Genes/DLX4ID49827ch17q21.html | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8 | |
| Rabbit | |
| 36 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 1748 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction