missing translation for 'onlineSavingsMsg'
Learn More

BP1 Antibody (1F11), Novus Biologicals™

Product Code. 18339077 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339077 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339077 Supplier Novus Biologicals Supplier No. H00001748M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BP1 Monoclonal antibody specifically detects BP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen BP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 1F11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH16145
Gene Alias Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8
Host Species Mouse
Immunogen DLX4 (AAH16145, 1 a.a. ~ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTSLPCPLPGRDASKAVFPDLAPVPSVAAAYPLGLSPTTAASPNLSYSRPYGHLLSYPYTEPANPGDSYLSCQQPAALSQPLCGPAEHPQELEADSEK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Growth and Development, Immunology, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1748
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.