missing translation for 'onlineSavingsMsg'
Learn More

BP-1-102, MedChemExpress

Product Code. 16495777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495777 5 mg 5mg
16489606 50 mg 50mg
16423048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495777 Supplier Medchem Express Supplier No. HY1004935mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BP-1-102 is an orally available, small-molecule inhibitor of transcription factor Stat3, with an IC50 of 6.8 μM.

BP-1-102 is an orally available, small-molecule inhibitor of transcription factor Stat3, with an IC50 of 6.8 μM.

Specifications

Chemical Name or Material BP-1-102
CAS 1334493-07-0
Color White
Purity Grade Notes Research
Molecular Formula C29H27F5N2O6S
Quantity 5 mg
Solubility Information DMSO : ≥ 33 mg/mL (52.67 mM)
SMILES O=C(O)C1=CC=C(N(CC2=CC=C(C3CCCCC3)C=C2)C(CN(C)S(=O)(C4=C(F)C(F)=C(F)C(F)=C4F)=O)=O)C=C1O
Molecular Weight (g/mol) 626.59
Formula Weight 626.59
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.