Learn More
Excel Healthcare Ltd™ Bowie Dick Test Pack
Product Code. 16340122
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 single use tests
Unit Size:
Each
Product Code. Quantity unitSize
16340122 10 single use tests Each
1 options
Product Code. 16340122 Supplier Excel Healthcare Ltd™ Supplier No. 189
This item requires a price update prior to being ordered.
Request a Price for this item here.
Detects atleast three vacuums are pulled per cycle
  • Excel Healthcare Has A Range Of Testing Products That Are Compatible With Our Range Of Steam Sterilisers
  • Manual Testing Of Steam Sterilisers Is Now Considered An Essential Requirement And Must Be Performed By The Operator On A Daily Basis
  • Htm 01-05 States That: Daily Tests Must Be Performed By The Operator Or User And If The Steriliser Fails To Meet Any Of The Test Requirements The Steriliser Must Be Withdrawn From Service And Advice Sought From The Manufacturer Or Service Provider
  • The Daily Tests Consist Of Steam Penetration Test (Vacuum Sterilisers Only) And Automatic Control Test. These Tests Should Be Dated, Signed And Recorded In The Steriliser Log Book
  • These Tests Are Easy To Complete And Record, Giving Full Performance Traceability Of Your Vacuum Steriliser
  • Efficient Testing Device
  • Detects Atleast Three Vacuums Are Pulled Per Cycle
Specifications
Certifications/Compliance Conform to EN IS0 1140 – 1 Class 2
Description Bowie Dick Test
Quantity 10 single use tests
Sterilization Indicator Test Steam: 134°C, 3, 5 min