missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Bovine Serum Albumin Native Protein

Product Code. 18461561 Shop All Bio Techne Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18461561

Brand: Novus Biologicals™ NBP19705810mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene ID (Entrez) 280717
Quantity 10 mg
Storage Requirements Store at 4°C.
Gene Alias albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbol ALB
Reconstitution Reconstitute with 1.0 ml deionized water.
Inhibitor Properties This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.
Research Category Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Purity or Quality Grade Multi-step
Protein Bovine Serum Albumin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.