missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Native Bovine Serum Albumin. Source: Bovine Serum The Bovine Serum Albumin Native Protein is derived from Bovine Serum. The Bovine Serum Albumin Native Protein has been validated for the following applications: Western Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
Specifications
Specifications
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000) |
| Gene ID (Entrez) | 280717 |
| Name | Bovine Serum Albumin protein |
| Storage Requirements | 4°C in the dark. |
| Cross Reactivity | Bovine |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?