missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Monoclonal Antibody (BGN/D1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182941
This item is not returnable.
View return policy
Description
This antibody does not react with goat, horse, human, mouse, rabbit or rat. Mouse anti Bovine serum albumin antibody, clone BGN/D1 recognizes bovine albumin, a major protein constituent of bovine serum.
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide | |
| P02769 | |
| ALB | |
| Bovine Serum Albumin. | |
| 1 mg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| BGN/D1 | |
| Unconjugated | |
| ALB | |
| ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine albumin; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; MGC179277; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
| Mouse | |
| Protein G | |
| RUO | |
| 280717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction