Learn More
Bovine Serum Albumin Antibody (D1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20114-0.025mg
 View more versions of this product
Product Code. 18628454
  • £190.00 / 0.02mg
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine samples. It is validated for ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bovine Serum Albumin
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Bovine
Purified
ELISA, Immunoprecipitation
D1
Unconjugated
albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
D1 was prepared by isolating anti-p24 scFvs from a phage display library made from a mouse immunized with lysate from HIV-1 infected H9 cells.
0.025 mg
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
280717
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ