missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (ALB/398), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45171-0.1MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in ELISA
Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) samples. It is validated for ELISA.
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| 66 kDa | |
| 0.1 mg | |
| Primary | |
| Reacts with a protein of ∼66kDa, identified as bovine serum albumin (BSA). It is a high affinity antibody and can be used for detection of traces of BSA. Bovine serum albumin (BSA) is an abundant plasma protein in cows that is important for maintaining osmotic pressure in blood plasma for proper distribution of body fluids between intravascular compartments and body tissues. BSA is a common buffer component for immunoglobulin type assays due to good solubility characteristics for water, Ca2+, Na+, K+, fatty acids, hormones and bilirubin. BSA makes up about half of the protein in plasma and represents the most stable and soluble protein in the plasma. It is a suitable reagent for laboratories developing immunoassays, mostly due to its availability, solubility and the numerous functional groups present for coupling. The BSA component contains several lysines that are capable of reacting with conjugation sites of linkers, making it applicable as a carrier protein for antigenic compounds. | |
| Store at 4C. | |
| IgG1 |
| ELISA | |
| ALB/398 | |
| Unconjugated | |
| 10mM PBS with 0.05% Sodium Azide | |
| ALB | |
| Purified bovine serum albumin | |
| Protein A or G purified | |
| RUO | |
| 280717 | |
| Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction