missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (3G3A2), Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | Bovine Serum Albumin |
|---|---|
| Clone | 3H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Simple Western, ELISA 1:10000 |
| Applications | Western Blot |
Description
Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Bovine samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
| Bovine Serum Albumin | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| ALB | |
| IgG1 | |
| Ascites | |
| Bovine serum albumin (BSA). |
| 3H6 | |
| Western Blot 1:500-1:2000, Simple Western, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine | |
| 280717 | |
| Bovine serum albumin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title