missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bovine Serum Albumin

Product Code. 16448768
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448768 10 g 10g
16412429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448768 Supplier MedChemExpress Supplier No. HYD0842
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bovine Serum Albumin (BSA) is a 583-residue protein consisting of three homologous all-α domains, organized in a heart-shaped structure. BSA is a globular protein that is used in numerous biochemical applications.

Specifications

Product Type Biochemical Assay Reagents
Quantity 10 g
Shipping Condition Room temperature
Storage Requirements 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Synonym Albumin bovine serum ∣ BSA
Target HBV
CAS 9048-46-8
Solubility H2O : 50 mg/mL (Need ultrasonic)
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.