missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Bovine Serum Albumin
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| Product Type | Biochemical Assay Reagents |
| Quantity | 1 g |
| Shipping Condition | Room temperature |
| Storage Requirements | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Synonym | Albumin bovine serum ∣ BSA |
| Target | HBV |
| CAS | 9048-46-8 |
| Solubility | H2O : 50 mg/mL (Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction