missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bovine FGF basic/FGF2/bFGF Protein
Description
The Bovine FGF basic/FGF2 Protein is derived from Bovine Brain. The Bovine FGF basic/FGF2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. |
| Gene ID (Entrez) | 281161 |
| Molecular Weight (g/mol) | 18 kDa, reducing conditions |
| Name | FGF basic |
| Quantity | 25 μg |
| Source | Bovine brain tissue-derived FGF basic/FGF2/bFGF protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?