missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Decorin ELISA Kit

Product Code. 17174779
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17174779

Brand: Invitrogen™ EB3RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Bovine Decorin quantitates bovine Decorin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant bovine Decorin.

Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product
Assay Range 5 to 1,200 pg/mL
Assay Sensitivity 5 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 280760
Gene Symbol DCN
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Decorin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.