missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Bovine Brain Extract (BBE), 9mg/mL, 5 mL

Product Code. 11615490
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11615490 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11615490 Supplier Lonza™ Supplier No. CC4098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful supplement in the culture of various endothelial cell types.

  • Suitable for use in the culture of various endothelial cell types.
  • Contains endothelial cell proliferation growth factors.
  • Available at a concentration of 9 mg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 9 mg/mL
Product Type Bovine Pituitary Extract
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.