missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bovine AGE-BSA Biotinylated Protein
Description
The Bovine AGE-BSA Biotinylated Protein is derived from Bovine Serum. The Bovine AGE-BSA Biotinylated Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Name | AGE-BSA |
| Quantity | 250 μg |
| Source | Bovine serum-derived AGE-BSA protein Advanced glycation end product (AGE) of bovine serum albumin (AGE-BSA) was prepared by incubating BSA and glucose under sterile conditions at 37 °C for 60 days and then biotinylated. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Biotin |
| Reconstitution | Reconstitute at 1 mg/mL in sterile PBS. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?