Learn More
Socorex™ Bottle Top Dispenser 530 Model
Brand:  Socorex™ 530.050
Code : NEW
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 13578290
  • £488.00 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Specifications
Bottletop Dispenser
EN ISO 8655
530
Autoclavable
5 to 50 mL