missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Bottle Rack
Shop All Agilent BioTek Products
Click to view available options
Holds:
Bottles
Unit Size:
Each
Description
- 8-bottle Rack for 25 mm (30 mL) Reagent Bottles
- For use with Agilent BioTek Microplate Washers and Readers
Specifications
Specifications
| Product Type | Bottle Rack |
| Holds | Bottles |
| Shelves | 6 |
| For Use With (Equipment) | For use with Agilent BioTek microplate washers and readers |
| Quantity | 1 Ea. |
| Diameter (Metric) Cavity | 25 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction