missing translation for 'onlineSavingsMsg'
Learn More

Interscience 2 L Bottle (GL 45)

Product Code. 12398727
Change view
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12398727 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12398727 Supplier Interscience Supplier No. 413002
  • ONLINE EXCLUSIVE PRICE
    £306.80
    £275.28 / Pack of 1
    Save £31.52
    10% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Set of 3 GL 45 glass bottle (2 L), autoclavable

Set of 3 GL 45 glass bottle (2 L), autoclavable

Specifications

Capacity (Metric) 2 L
Description GL45 glass bottle (2 L), autoclavable
For Use With (Equipment) easySpiral
Autoclavable Autoclavable
Product Type Reagent Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.