missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ Bottle Cap Assembly with 3 Holes
Click to view available options
Color: Red; 2 x 1/8 in. and 1 x 3/16 in. holes
Specifications
For Use With (Equipment) | GL-45 thread, 1mL bottles, 1 x 3/16 in. waters type tubing and 2 x 1/8 in. standard tubing |
Product Type | 3 Hole Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'