missing translation for 'onlineSavingsMsg'
Learn More

Bosutinib, MedChemExpress

Product Code. 16420936
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16420936

Brand: Medchem Express HY10158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bosutinib is an oral Src/Abl tyrosine kinase inhibito with IC50 of 1.2 nM and 1 nM, respectively.

Bosutinib is an oral Src/Abl tyrosine kinase inhibito with IC50 of 1.2 nM and 1 nM, respectively.

Specifications

Chemical Name or Material Bosutinib
CAS 380843-75-4
Color White
Purity Grade Notes Research
Molecular Formula C26H29Cl2N5O3
Quantity 200 mg
Synonym SKI-606
Solubility Information DMSO : ≥ 46 mg/mL (86.72 mM)
SMILES CN1CCN(CC1)CCCOC2=C(C=C3C(NC4=CC(OC)=C(C=C4Cl)Cl)=C(C=NC3=C2)C#N)OC
Molecular Weight (g/mol) 530.45
Formula Weight 530.45
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.