missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Boston S1P Shoe

Product Code. p-7225069
Change view
Click to view available options
Size:
35
36
37
38
48
Unit Size:
1 pair
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17572456 35 1 pair
17582456 36 1 pair
17592456 37 1 pair
17502466 38 1 pair
17512466 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17572456 Supplier Emma Safety Footwear Supplier No. MM40064735
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Extra light, extra strong. The Boston safety shoe combines optimal comfort with optimal protection.

  • Extra light, extra strong. This Athletic safety shoe combines optimal comfort with optimal protection. And it's got the looks too. Thanks to the knitted textile upper and the Nano Carbon Composite protective nose, the Boston is particularly light and comfortable. Ideal for workplaces where safety and long-lasting comfort are top priorities.
  • The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable. The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ Antibacterial lining provides optimal breathability. Boston exudes pure sportsmanship and is designed for performance and looks.
  • Industries
    • Logistics & transport
    • Electronics
    • Manufacturing
  • Safety shoe
    • S1P-Safetyshoe with sole plate for dry work

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Material Polyurethane
Size 35
Toe Material Composite
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.