missing translation for 'onlineSavingsMsg'
Learn More

Bosentan USP Related Compound D, Mikromol™

Product Code. 30052678
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052678 25 mg 25mg
17983576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052678 Supplier Mikromol Supplier No. MM3369.030025
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bosentan USP Related Compound D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
CAS 150728-13-5
Analyte or Component Names 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
Shipping Condition Room Temperature
Molecular Formula C15 H10 Cl2 N4 O2
Certifications/Compliance ISO 17025
Synonym 2,2'-Bipyrimidine, 4,6-dichloro-5-(2-methoxyphenoxy)-, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-dipyrimidine, 4,6-Dichloro-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)pyrimidine, (4,6-dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine), Bosentan Related Compound D, Bosentan Related Compound D
InChI Formula InChI=1S/C15H10Cl2N4O2/c1-22-9-5-2-3-6-10(9)23-11-12(16)20-15(21-13(11)17)14-18-7-4-8-19-14/h2-8H,1H3
SMILES COc1ccccc1Oc2c(Cl)nc(nc2Cl)c3ncccn3
IUPAC Name 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
Molecular Weight (g/mol) 349.17
Quantity 25 mg
Formula Weight 348.0181 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.