missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bosentan

Product Code. 18758606
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758606 10 mg 10mg
18706158 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18758606 Supplier Tocris Bioscience™ Supplier No. 6232/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity dual ETA and ETB receptor antagonist;orally bioavailable

Bosentan is a high affinity dual ETA and ETB receptor antagonist (Ki values are 4.7 and 95 nM for ETA and ETB, respectively). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C -induced contraction of rat trachea in vitro. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

CAS 147536-97-8
Quantity 10 mg
Target Non-selective Endothelin Receptor
Molecular Formula C27H29N5O6S
Formula Weight 551.61g/mol
Product Type Antagonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.