missing translation for 'onlineSavingsMsg'
Learn More

Bosentan (hydrate), MedChemExpress

Product Code. 16491346
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
50mg
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491346 200 mg 200mg
16481999 1 g 1g
16474147 50 mg 50mg
16457986 100 mg 100mg
16401976 500 mg 500mg
16400708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16491346 Supplier Medchem Express Supplier No. HYA0013A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors with Ki of 4.7 nM and 95 nM in human SMC, respectively.

Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors with Ki of 4.7 nM and 95 nM in human SMC, respectively.

Specifications

Chemical Name or Material Bosentan (hydrate)
CAS 157212-55-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H31N5O7S
Quantity 200 mg
Solubility Information DMSO : ≥ 100 mg/mL (175.55 mM) ∣Ethanol : 50 mg/mL (87.78 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(NC1=NC(C2=NC=CC=N2)=NC(OCCO)=C1OC3=CC=CC=C3OC)(C4=CC=C(C(C)(C)C)C=C4)=O.O
Molecular Weight (g/mol) 569.63
Formula Weight 569.63
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.