missing translation for 'onlineSavingsMsg'
Learn More

Bortezomib, MedChemExpress

Product Code. 16476457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476457 5 mg 5mg
16444796 100 mg 100mg
16413728 10 mg 10mg
16470107 50 mg 50mg
16470956 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476457 Supplier Medchem Express Supplier No. HY10227
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bortezomib (PS-341) is a reversible and selective proteasome inhibitor, and potently inhibits 20S proteasome (Ki=0.6 nM) by targeting a threonine residue. Bortezomib disrupts the cell cycle, induces apoptosis, and inhibits NF-κB. Bortezomib is the first proteasome inhibitor anticancer agent. Anti-cancer activity.

Bortezomib (PS-341) is a reversible and selective proteasome inhibitor, and potently inhibits 20S proteasome (Ki=0.6 nM) by targeting a threonine residue. Bortezomib disrupts the cell cycle, induces apoptosis, and inhibits NF-κB. Bortezomib is the first proteasome inhibitor anticancer agent. Anti-cancer activity.

Specifications

Chemical Name or Material Bortezomib
CAS 179324-69-7
Color White
Purity Grade Notes Research
Molecular Formula C19H25BN4O4
Quantity 5 mg
Synonym PS-341 LDP-341 NSC 681239
Solubility Information Ethanol : 66.67 mg/mL (173.51 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 50 mg/mL (130.13 mM; Need ultrasonic)
SMILES OB(O)[C@H](CC(C)C)NC([C@@H](NC(C1=NC=CN=C1)=O)CC2=CC=CC=C2)=O
Molecular Weight (g/mol) 384.24
Formula Weight 384.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.