missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bortezomib
GREENER_CHOICE

Product Code. 18788286
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18788286

Brand: Tocris Bioscience™ 7282/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity proteasome inhibitor

Bortezomib is a high affinity proteasome inhibitor (Ki = 0.6 nM). Inhibits growth of multiple myeloma (MM) cell lines (IC50 values in <50 nM range); induces apoptosis in MM cell lines and MM patient derived cells, including p53 mutant cell lines. Decreases adherence of MM cells lines to bone marrow stromal cells (BMSCs). Acts synergistically with dexamethasone (Cat. No. 1126). Also cytotoxic in MCF-7 breast cancer cell line (IC50 = 50 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material B-[(1 R)-3-Methyl-1-[[(2 S)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]boronic acid
CAS 179324-69-7
Quantity 25 mg
Target Proteasome Inhibitors
Molecular Formula C19H25BN4O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.