missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Glass Weighing Scoops

Product Code. 10064453
Change view
Click to view available options
Length (Metric):
82 mm
95 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10064453 82 mm Each
10465205 95 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10064453 Supplier Lenz Laborglasinstrumente™ Supplier No. 05568003
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Empties completely without any loss of material. Lenz Laborglasinstrumente™ Glass Weighing Scoops are manufactured from durable DURAN™ borosilicate glass.

  • Weighing scoop/funnel combination
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock

Specifications

Quantity 1 Ea.
Capacity (Metric) 3 mL
Product Type Weighing Scoop
For Use With (Application) Used for complete emptying without losses
Material Borosilicate Glass
Length (Metric) 82 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.