missing translation for 'onlineSavingsMsg'
Learn More

Kimble™ TITESEAL™ Glass Vials

Product Code. 11863183
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 144
Product Code. Color unitSize
11863183 Clear Pack of 144
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 11863183

Brand: Kimble™ 60965D3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

An excellent vial option for the laboratory with open-bottom plug style unattached plastic closures.

  • Plain tops
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
  • Provided in a 144 pack
TRUSTED_SUSTAINABILITY

Specifications

Classification A
Color Clear
Diameter (Metric) Outer 19 mm
Material Borosilicate Glass
Volume (Metric) 11 mL
No. per Pack 144
Closure Material Plastic
Closure Type Titeseal
Quantity 144 Pk.
Height (Metric) 65 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.