Learn More
Torion Glass Body with Fittings
Description
- Only glass and PTFE come into contact with the liquid being used to provide a chemical resistant pathway
- Complete joint comprises borosilicate glass body, glass filled polypropylene fitting and rubber/PTFE sealing ring
- Glass body has an internal screwthread into which is fitted the rubber sealing ring sheathed with PTFE
Specifications
Specifications
| Material | Borosilicate Glass |
| For Use With | Glassblowers jointing system |
| Height (Metric) | 32 mm |
| Product Type | Torion Glass Body |