Learn More
Borosilicate Glass T-shape Stopcock Adapter
Control flow rate with a T-shape adapter. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand:  Quickfit MF11/3B/SC
79.52 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.04324kg
 View more versions of this product
Product Code. 10349804
  • £178.00 / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
  • Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Specifications
Stopcock Adapter
24/29
GP Rotaflo™ PTFE stopcock
Borosilicate Glass
T-shape