Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Click to view available options
Port Size:
14/23
19/26
24/29
Unit Size:
Each
Pack of 5
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Specifications
| Material | Borosilicate Glass |
| Port Size | 19/26 |
| Includes | Sockets |
| Product Type | Stopcock Adapter |