Learn More
Borosilicate Glass Stopcock Adapter with Cone
Control flow rate. Quickfit Jointed Ware™ QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
£83.20 - £135.00
Specifications
Product Type | Stopcock Adapter |
---|---|
Material | Borosilicate Glass |
Includes | Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
11382035
|
Quickfit
MF11/1/SC |
14/23 |
£135.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
11302045
|
Quickfit
MF11/2/SC |
19/26 |
£123.00
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11322045
|
Quickfit
MF11/3/SC |
24/29 |
£130.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
11332045
|
Quickfit
MF11/4/SC |
29/32 |
£83.20
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
Control flow rate. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
- With GP Rotaflo™ PTFE stopcock: - No lubrication required - No contaminating greases - Fine flow control - Positive closure
- Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Stopcock Adapter | |
Borosilicate Glass | |
Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock |