missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Borosilicate Glass Serological Pipette
Description
- Ideal for use with radioactive and other hazardous materials
- Volumes are color-coded
- Environmentally friendly compared with plastic counterparts
Specifications
Specifications
| Material | Borosilicate Glass |
| Graduated | Yes |
| Graduations | 0.1 mL |
| Volume (Metric) | 10 mL |
| Length (Metric) | 300 mm |
| Type | Serological Pipette |
| Disposable | Single-use |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?