Learn More
Day Impex™ Borosilicate Glass Refill
Designed with practicality and safety in mind. Day Impex™ Borosilicate Glass Refill is ultra-low evacuated to allow for safe and efficient handling of all liquified gases, freezing mixtures, and specimens.
Brand: Day Impex™ 210007
Code : 03
Additional Details : Weight : 0.80000kg
Description
- Borosilicate glass inner vessels
Specifications
90 mm | |
For use with Enamelled Steel container | |
240 mm | |
1500 mL |
4.5 in. | |
240mm | |
Borosilicate Glass |