Learn More
Hamilton™ Terasaki Dispenser
Product Code. 11567944
Change view
Click to view available options
Capacity (Metric):
25 μL
500 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11567944 500 μL Each
11547914 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11567944 Supplier Hamilton™ Supplier No. 201940
Only null left
Add to basket
Add to basket
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
  • Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
  • Uses removable needles
  • Available for a range of volumes
Specifications
Quantity 1/Ea.
Type Gastight Syringe
For Use With (Application) Liquid or gas sampling
For Use With (Equipment) Model 1702 Terasaki Dispenser
Material PTFE (Plunger)
Capacity (Metric) 500 μL