Learn More
Hamilton™ Terasaki Dispenser
Description
- Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
- Uses removable needles
- Available for a range of volumes
Specifications
Specifications
| Quantity | 1/Ea. |
| Type | Gastight Syringe |
| For Use With (Application) | Liquid or gas sampling |
| For Use With (Equipment) | Model 1702 Terasaki Dispenser |
| Material | PTFE (Plunger) |
| Capacity (Metric) | 25 μL |