missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Borosilicate Glass Pasteur Pipets

Product Code. 10494533 Shop All Corning Products
Change view
Click to view available options
Length (Metric):
146 mm
229 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10494533 146 mm Pack of 1000
10323523 229 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10494533 Supplier Corning™ Supplier No. 7095D5X
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured with Borosilicate Glass for optimum visibility. Corning™ Borosilicate Glass Pasteur Pipets are ideal for rapid non-volumetric transfer work.

  • Available in convenient bulk-packs
  • One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Feature a constriction one inch below the top to facilitate plugging
  • Ideal for work in bacteriology, immunology, hematology and serology studies
  • Use for blood bank and clinical chemistry procedures
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 6.5 mm
Material Borosilicate Glass
For Use With (Application) Ideal for rapid non-volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures
Quantity 1000 Pack
Tip Style Round
Length (Metric) 146 mm
Type Pasteur Pipet
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.