Learn More
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01400334
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11876164
  • £54.43 / Pack of 5
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow stopper
5/Pk.
Borosilicate Glass
NS 34/35