Learn More
Quickfit™ Borosilicate Glass Distillation Head
Description
- With integral jacketed condenser
- Liquid leaving condenser is directed towards a GP Rotaflo™ stopcock
- Adjustment of stopcock controls proportion of liquid returned to the column or collected
- Screwcap fitting for insertion of thermometer
- Side-arm for venting or connection to vacuum supply
Specifications
Specifications
| Type | Distillation Head |
| Material | Borosilicate Glass |
| Certifications/Compliance | ISO 3585 |
| For Use With (Equipment) | Integral jacketed condenser |
| Standard Taper Joint | 29/32, 19/26 |