Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04000037
Additional Details : Weight : 0.11100kg
 View more versions of this product
Product Code. 10728731
  • £76.00 / Pack of 2
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
Borosilicate Glass
2Pack
Polyethylene