Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Solid or semi-hollow
- Octagonal grip
Specifications
Specifications
| Quantity | 1/Pk. |
| Standard Taper Joint | NS 29/32 |
| Material | Borosilicate Glass |
| Product Type | Ground Joint Stopper |
| Shape | Octagonal Grip (Semi-hollow) |