missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Borosilicate Glass Class A Mortar

Brand:  Wheaton™ 358136

 View more versions of this product

Product Code. 11778272

  • £141.00 / Pack of 2
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
3 mL
Mortar
86mm
11mm
Borosilicate Glass
For 358107 (3mL) Tapered Tissue Grinder Set
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers