missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Borosilicate Glass Class AS Burette with Teflon Tap

Complies with BS-IS 385 standards. Fisherbrand™ Borosilicate Glass Class AS Burette with Teflon Tap includes single bore with interchangeable PTFE stopcock. Blue ceramic graduations make volume assessment easy.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.30600kg

 View more versions of this product

Product Code. 11566913

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Constructed of borosilicate glass
  • Single bore with interchangeable PTFE stopcock
  • Blue ceramic graduations
  • Class AS, BS-ISO 385 Compliance
Specifications

Specifications

50 mL
0.1 mL
Blue
±0.05 mL
Class AS
Borosilicate Glass
Single Bore, Interchangeable Stopcock
PTFE
Buret
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers