Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand: BRAND™ 144620
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Hollow
- Hexagonal grip
Specifications
Conical Ground Joint Stopper | |
1/Pk. | |
NS 14/23 |
NS 14/23 | |
Borosilicate Glass |