Learn More
Lenz Laborglasinstrumente™ Borosilicate Drying Tubes
Product Code. 10326311
Change view
Click to view available options
Shape:
Straight
U-shaped
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shape Unit Size
10326311 Straight Each
10264241 U-shaped Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10326311 Supplier Lenz Laborglasinstrumente™ Supplier No. 05330010
  • ONLINE EXCLUSIVE PRICE
    £9.00
    £6.60 / Pack of 1
    Save £2.40
    26% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Diameter (Metric) Outer 13 mm
Length (Metric) 100 mm
Shape Straight
Material Borosilicate Glass
Product Type Drying Tube