missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride etherate, approx. 48% BF3

Product Code. 10593921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
2.5kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10593921 2.5 kg Glass bottle 2.5kg
10042110 25 g Glass bottle 25g
10296130 100 g Glass bottle 100g
10286130 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10593921 Supplier Thermo Scientific Acros Supplier No. 174560025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
Synonym Boron trifluoride ethyl ether
PubChem CID 8000
IUPAC Name ethoxyethane;trifluoroborane
SMILES FB(F)F.CCOCC

Specifications

Melting Point -60.0°C
Color Colorless to Straw-Yellow
Density 1.1200g/mL
Boiling Point 126.0°C
Flash Point 58°C
Packaging Glass bottle
Linear Formula (C2H5)2O·BF3
Refractive Index 1.3429 to 1.3449
Quantity 2.5 kg
Fieser 01,70; 02,35; 03,33; 04,44; 05,52; 06,65; 07,31; 08,51; 09,64; 10,52; 11,72; 12,66; 13,3; 14,54; 15,45; 16,44; 17,52
Merck Index 15,1352
Specific Gravity 1.12
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether
Viscosity 1.89 mPa.s (20°C)
Formula Weight 141.93
Percent Purity ∼48% BF3
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Boron trifluoride etherate, 48% BF3
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.