missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Melting Point | -14.0°C |
| Color | Brown |
| Density | 1.2300g/mL |
| Boiling Point | 126.0°C to 127.0°C |
| Flash Point | 62°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Linear Formula | (CH3)2O·BF3 |
| Refractive Index | 1.3070 to 1.3090 |
| Quantity | 250 mL |
| Show More |
Safety and Handling
Product Identifier
- Boron trifluoride dimethyl etherate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?