missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. p-7058662
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330258 100 mL 100mL
11396169 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330258 Supplier Thermo Scientific Alfa Aesar Supplier No. 042970.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-43-5
Molecular Formula C2H6BF3S
Molecular Weight (g/mol) 129.94
MDL Number MFCD00013193
InChI Key BRWZPVRDOUWXKE-UHFFFAOYSA-N
Synonym boron fluoride-dimethyl sulfide complex, dimethyl-??-sulfanylidene trifluoro-??-borane, trifluoroborane-methyl sulfide, dimethyl sulfide-trifluoroborane, trifluoro methylsulfanyl methane boron, boron trifluoride methyl sulfide complex, boron trifluoride-methyl sulfide complex, boron trifluoride-dimethyl sulfide complex, purified, packaged under argon in resealable chemseal bottles
PubChem CID 71311438
IUPAC Name dimethylsulfonio(trifluoro)boranuide
SMILES CSC.FB(F)F

Specifications

Physical Form Liquid
Quantity 100 mL
UN Number UN2920
Formula Weight 129.94
Packaging ChemSeal™ bottles
Sensitivity Moisture sensitive
Density 1.235 g/mL
Chemical Name or Material Boron trifluoride-dimethyl sulfide complex, Purified

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.